Search
Search
| Gene Symbol: Dmp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006250622.2 | XP_006250684.1 | 7 - 8 | 2720 | 77 | |
| XM_008769994.1 | XP_008768216.1 | 3 - 4 | 418 | 77 | ||
| XM_008769995.1 | XP_008768217.1 | 6 - 7 | 837 | 77 | ||
Set Membership: |
|