Search
Search
| Gene Symbol: Atp11c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006257596.2 | XP_006257658.1 | 8 - 9 | 720 | 97 | |
| XM_006257597.2 | XP_006257659.1 | 8 - 9 | 1733 | 97 | ||
| XM_006257599.2 | XP_006257661.1 | 8 - 9 | 1733 | 97 | ||
Set Membership: |
|