Search
Search
| Gene Symbol: Asgr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012503.2 | NP_036635.1 | 7 - 8 | 651 | 97 | |
| XM_006246579.1 | XP_006246641.1 | 7 - 8 | 763 | 97 | ||
| XM_008767768.1 | XP_008765990.1 | 7 - 8 | 783 | 97 | ||
| XM_008767769.1 | XP_008765991.1 | 7 - 8 | 727 | 97 | ||
| GenBank mRNA | BC088154.1 | - | 7 - 8 | 651 | 7372227 | 97 |
| FQ209763.1 | - | 7 - 8 | 687 | 97 | ||
| FQ218553.1 | - | 7 - 8 | 727 | 97 | ||
| FQ218644.1 | - | 7 - 8 | 687 | 97 | ||
| FQ218824.1 | - | 7 - 8 | 687 | 97 | ||
| FQ219107.1 | - | 7 - 8 | 724 | 97 | ||
| FQ219710.1 | - | 7 - 8 | 768 | 97 | ||
| M21770.1 | - | 6 - 7 | 553 | 97 | ||
Set Membership: |
|