Search
Search
| Gene Symbol: Snf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007804.1 | NP_001007805.1 | 3 - 4 | 243 | 72 | |
| XM_006247185.1 | XP_006247247.1 | 3 - 4 | 376 | 72 | ||
| GenBank mRNA | BC086364.1 | - | 3 - 4 | 286 | 7304427 | 72 |
Set Membership: |
|