Search
Search
| Gene Symbol: Phc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107886.1 | NP_001101356.1 | 7 - 8 | 1232 | 107 | |
| XM_006237300.2 | XP_006237362.1 | 8 - 9 | 1257 | 107 | ||
| XM_006237302.2 | XP_006237364.1 | 6 - 7 | 1155 | 107 | ||
| XM_006237303.2 | XP_006237365.1 | 7 - 8 | 1097 | 107 | ||
Set Membership: |
|