Search
Search
| Gene Symbol: Clec9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109354.1 | NP_001102824.1 | 2 - 3 | 274 | 72 | |
| XM_006237099.2 | XP_006237161.1 | 2 - 3 | 1150 | 72 | ||
| XM_008763325.1 | XP_008761547.1 | 1 - 2 | 452 | 72 | ||
| XM_008763326.1 | XP_008761548.1 | 2 - 3 | 151 | 72 | ||
| GenBank mRNA | GU357486.1 | - | 2 - 3 | 178 | 72 | |
Set Membership: |
|