Search
Search

LOC100910210
LOC103690013
Nhp2| Gene Symbol: LOC100910210 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | FQ222023.1 | - | 1 - 2 | 218 | 113 | |
| Gene Symbol: LOC103690013 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006246240.2 | XP_006246302.1 | 1 - 2 | 237 | 113 | |
| Gene Symbol: Nhp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003750818.3 | XP_003750866.1 | 1 - 2 | 237 | 113 | |
| GenBank mRNA | BC162010.1 | - | 1 - 2 | 176 | 9091322 | 113 |
Set Membership: |
|