Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030872.1 | NP_110499.1 | 1 - 2 | 216 | 133 | |
GenBank mRNA | BC061823.1 | - | 1 - 2 | 180 | 5599188 | 133 |
U10357.1 | - | 1 - 2 | 216 | 133 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Rattus norvegicus Probe spans exons |