Search
Search
| Gene Symbol: Tbcd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271364.1 | NP_001258293.1 | 35 - 36 | 3339 | 60 | |
| XM_006247861.2 | XP_006247923.1 | 35 - 36 | 3400 | 60 | ||
| XM_006247864.2 | XP_006247926.1 | 35 - 36 | 3400 | 60 | ||
| XM_008768500.1 | XP_008766722.1 | 33 - 34 | 3316 | 60 | ||
| XM_008768501.1 | XP_008766723.1 | 36 - 37 | 3402 | 60 | ||
| XM_008768502.1 | XP_008766724.1 | 31 - 32 | 2807 | 60 | ||
Set Membership: |
|