Search
Search
| Gene Symbol: Rfwd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025126.1 | NP_001020297.1 | 8 - 9 | 1016 | 100 | |
| XM_006250113.2 | XP_006250175.1 | 15 - 16 | 2215 | 100 | ||
| XM_006250114.2 | XP_006250176.1 | 15 - 16 | 2203 | 100 | ||
| XM_006250115.2 | XP_006250177.1 | 14 - 15 | 2155 | 100 | ||
| XM_006250116.2 | XP_006250178.1 | 14 - 15 | 2143 | 100 | ||
| GenBank mRNA | BC091284.1 | - | 8 - 9 | 1016 | 7304516 | 100 |
Set Membership: |
|