Search
Search
| Gene Symbol: Plk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031821.1 | NP_114009.1 | 5 - 6 | 833 | 98 | |
| GenBank mRNA | AF136583.1 | - | 5 - 6 | 833 | 98 | |
| BC070878.1 | - | 5 - 6 | 817 | 7096750 | 98 | |
Set Membership: |
|