Search
Search
| Gene Symbol: Dmxl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003751784.3 | XP_003751832.1 | 19 - 20 | 5066 | 64 | |
| XM_006254710.2 | XP_006254772.1 | 19 - 20 | 5066 | 64 | ||
| XM_006254711.2 | XP_006254773.1 | 19 - 20 | 5066 | 64 | ||
| XM_006254712.2 | XP_006254774.1 | 19 - 20 | 5066 | 64 | ||
Set Membership: |
|