Search
Search
| Gene Symbol: Cdca5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001115042.2 | NP_001108514.2 | 8 - 9 | 1067 | 103 | |
| XM_008760177.1 | XP_008758399.1 | 4 - 5 | 727 | 103 | ||
| GenBank mRNA | BC097411.1 | - | 8 - 9 | 997 | 103 | |
| BC158663.1 | - | 8 - 9 | 1067 | 8369770 | 103 | |
| BC158764.1 | - | 8 - 9 | 1006 | 7111047 | 103 | |
Set Membership: |
|