Search
Search
| Gene Symbol: Ntm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017354.1 | NP_059050.1 | 5 - 6 | 1266 | 128 | |
| XM_006242749.1 | XP_006242811.1 | 5 - 6 | 1584 | 128 | ||
| XM_008766065.1 | XP_008764287.1 | 5 - 6 | 1584 | 128 | ||
| GenBank mRNA | U16845.1 | - | 5 - 6 | 1266 | 128 | |
Set Membership: |
|