Search
Search
| Gene Symbol: Cyp3a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_147206.2 | NP_671739.2 | 3 - 4 | 310 | 78 | |
| GenBank mRNA | BC086985.1 | - | 3 - 4 | 310 | 7125835 | 78 |
| U60085.2 | - | 3 - 4 | 286 | 78 | ||
Set Membership: |
|