Search
Search
| Gene Symbol: Dcx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053379.3 | NP_445831.3 | 5 - 6 | 1012 | 91 | |
| XM_006257382.2 | XP_006257444.1 | 5 - 6 | 1137 | 91 | ||
| XM_006257385.2 | XP_006257447.1 | 5 - 6 | 1230 | 91 | ||
| GenBank mRNA | AF155959.2 | - | 4 - 5 | 946 | 91 | |
Set Membership: |
|