Search
Search

Slc17a4| Gene Symbol: Slc17a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271214.1 | NP_001258143.1 | 7 - 8 | 1018 | 74 | |
| XM_008771657.1 | XP_008769879.1 | 7 - 8 | 1023 | 74 | ||
| XM_008771658.1 | XP_008769880.1 | 8 - 9 | 1337 | 74 | ||
| XM_008771659.1 | XP_008769881.1 | 8 - 9 | 1407 | 74 | ||
| XM_008771660.1 | XP_008769882.1 | 6 - 7 | 789 | 74 | ||
Set Membership: |
|