Search
Search
| Gene Symbol: Celf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109190.2 | NP_001102660.1 | 3 - 4 | 1045 | 58 | |
| XM_006232897.2 | XP_006232959.1 | 3 - 4 | 971 | 58 | ||
| XM_006232902.2 | XP_006232964.1 | 3 - 4 | 971 | 58 | ||
| XM_006232903.2 | XP_006232965.1 | 2 - 3 | 1342 | 58 | ||
| XM_006232904.2 | XP_006232966.1 | 4 - 5 | 2699 | 58 | ||
| XM_006232905.2 | XP_006232967.1 | 4 - 5 | 2732 | 58 | ||
| XM_006232906.2 | XP_006232968.1 | 2 - 3 | 3804 | 58 | ||
| XM_006232907.2 | XP_006232969.1 | 2 - 3 | 289 | 58 | ||
| XM_006232908.2 | XP_006232970.1 | 3 - 4 | 969 | 58 | ||
| XM_008761322.1 | XP_008759544.1 | 4 - 5 | 3286 | 58 | ||
| XM_008761323.1 | XP_008759545.1 | 3 - 4 | 255 | 58 | ||
| XM_008761324.1 | XP_008759546.1 | 2 - 3 | 2409 | 58 | ||
| XM_008761325.1 | XP_008759547.1 | 2 - 3 | 2363 | 58 | ||
Set Membership: |
|