Search
Search
| Gene Symbol: Hmgcs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173094.2 | NP_775117.2 | 7 - 8 | 1324 | 93 | |
| XM_006233002.2 | XP_006233064.1 | 7 - 8 | 1353 | 93 | ||
| GenBank mRNA | BC078695.1 | - | 7 - 8 | 1324 | 7112644 | 93 |
| BC083543.1 | - | 7 - 8 | 1324 | 7132009 | 93 | |
| M33648.1 | - | 7 - 8 | 1342 | 93 | ||
Set Membership: |
|