Search
Search

Hormad1| Gene Symbol: Hormad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108949.1 | NP_001102419.1 | 7 - 8 | 407 | 139 | |
| XM_006232951.2 | XP_006233013.1 | 6 - 7 | 362 | 139 | ||
Set Membership: |
|