Search
Search
| Gene Symbol: Eno1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109908.1 | NP_001103378.1 | 6 - 7 | 611 | 60 | |
| NM_012554.3 | NP_036686.2 | 6 - 7 | 562 | 60 | ||
| XM_006239444.1 | XP_006239506.1 | 6 - 7 | 578 | 60 | ||
| GenBank mRNA | BC063174.1 | - | 6 - 7 | 555 | 60 | |
| BC078896.1 | - | 6 - 7 | 611 | 7112523 | 60 | |
| BC081847.1 | - | 6 - 7 | 535 | 7189453 | 60 | |
| BC090069.1 | - | 6 - 7 | 492 | 7108936 | 60 | |
| BC091572.1 | - | 6 - 7 | 532 | 7107492 | 60 | |
| BC097343.1 | - | 6 - 7 | 528 | 7461555 | 60 | |
| FQ214707.1 | - | 6 - 7 | 484 | 60 | ||
| FQ225935.1 | - | 6 - 7 | 575 | 60 | ||
| X02610.1 | - | 6 - 7 | 60 | |||
Set Membership: |
|