Search
Search

Rad51
LOC100911267| Gene Symbol: Rad51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109204.1 | NP_001102674.1 | 8 - 9 | 995 | 82 | |
| XM_006234784.2 | XP_006234846.1 | 9 - 10 | 1173 | 82 | ||
| GenBank mRNA | BC168875.1 | - | 8 - 9 | 959 | 9022295 | 82 |
| Gene Symbol: LOC100911267 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008762135.1 | XP_008760357.1 | 4 - 5 | 532 | 82 | |
Set Membership: |
|