Search
Search

Atxn7l2| Gene Symbol: Atxn7l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006233199.2 | XP_006233261.1 | 6 - 7 | 900 | 96 | |
| XM_006233200.2 | XP_006233262.1 | 6 - 7 | 900 | 96 | ||
| XM_006233204.2 | XP_006233266.1 | 6 - 7 | 2263 | 96 | ||
| XM_008761446.1 | XP_008759668.1 | 7 - 8 | 1023 | 96 | ||
| GenBank mRNA | BC098000.1 | - | 6 - 7 | 779 | 96 | |
Set Membership: |
|