Search
Search
| Gene Symbol: Lrrc48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013857.1 | NP_001013879.1 | 2 - 3 | 285 | 99 | |
| XM_006246462.2 | XP_006246524.1 | 3 - 4 | 553 | 99 | ||
| XM_006246463.2 | XP_006246525.1 | 3 - 4 | 332 | 99 | ||
| XM_006246464.2 | XP_006246526.1 | 3 - 4 | 884 | 99 | ||
| XM_006246465.2 | XP_006246527.1 | 3 - 4 | 445 | 99 | ||
| XM_008767782.1 | XP_008766004.1 | 4 - 5 | 439 | 99 | ||
| XM_008767783.1 | XP_008766005.1 | 3 - 4 | 1649 | 99 | ||
| XM_008767784.1 | XP_008766006.1 | 4 - 5 | 1925 | 99 | ||
| XM_008767785.1 | XP_008766007.1 | 3 - 4 | 332 | 99 | ||
| GenBank mRNA | BC083838.1 | - | 2 - 3 | 285 | 7113397 | 99 |
Set Membership: |
|