Search
Search
| Gene Symbol: Gpr64 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270872.1 | NP_001257801.1 | 4 - 5 | 255 | 145 | |
| XM_008773156.1 | XP_008771378.1 | 4 - 5 | 683 | 145 | ||
| GenBank mRNA | AF538959.1 | - | 4 - 5 | 265 | 145 | |
Set Membership: |
|