Search
Search
| Gene Symbol: Syt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033680.2 | NP_001028852.2 | 4 - 5 | 505 | 85 | |
| XM_008765352.1 | XP_008763574.1 | 1 - 2 | 487 | 85 | ||
| XM_008765353.1 | XP_008763575.1 | 1 - 2 | 490 | 85 | ||
| GenBank mRNA | AA924659.1 | - | 3 - 4 | 85 | ||
| AJ617617.1 | - | 3 - 4 | 404 | 85 | ||
| AJ617620.1 | - | 4 - 5 | 625 | 85 | ||
| AJ617621.1 | - | 4 - 5 | 505 | 85 | ||
| AJ617622.1 | - | 3 - 4 | 326 | 85 | ||
| DQ181550.2 | - | 1 - 2 | 173 | 85 | ||
| X52772.1 | - | 4 - 5 | 688 | 85 | ||
Set Membership: |
|