Search
Search
| Gene Symbol: Tmsb4x | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031136.1 | NP_112398.1 | 3 | 481 | 76 | |
| GenBank mRNA | BC058137.1 | - | 3 | 495 | 6889807 | 76 |
| FQ211950.1 | - | 3 | 491 | 76 | ||
| FQ229728.1 | - | 3 | 489 | 76 | ||
| M34043.1 | - | 3 | 481 | 76 | ||
Set Membership: |
|