Search
Search

ATXN7L1| Gene Symbol: ATXN7L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003482699.4 | XP_003482747.1 | 1301 | 61 | ||
| XM_003482700.4 | XP_003482748.1 | 1331 | 61 | |||
| XM_005667723.3 | XP_005667780.1 | 1004 | 61 | |||
| XM_005667724.3 | XP_005667781.1 | 1332 | 61 | |||
| XM_021063503.1 | XP_020919162.1 | 1331 | 61 | |||
| XM_021063504.1 | XP_020919163.1 | 744 | 61 | |||
| XM_021063505.1 | XP_020919164.1 | 1334 | 61 | |||
| XM_021063506.1 | XP_020919165.1 | 1331 | 61 | |||
| XM_021063513.1 | XP_020919172.1 | 285 | 61 | |||
| GenBank mRNA | AK237402.1 | - | 1 - 2 | 115 | 61 | |
| AK394227.1 | - | 1 - 2 | 114 | 61 | ||
| AK400708.1 | - | 1 - 2 | 169 | 61 | ||
Set Membership: |
|