Search
Search
| Gene Symbol: WDR48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003132119.4 | XP_003132167.2 | 518 | 122 | ||
| XM_013981475.2 | XP_013836929.2 | 371 | 122 | |||
| XM_013981476.2 | XP_013836930.1 | 518 | 122 | |||
| GenBank mRNA | AK230485.1 | - | 4 - 5 | 376 | 122 | |
| AK348040.1 | - | 5 - 6 | 489 | 122 | ||
Set Membership: |
|