Search
Search
| Gene Symbol: RORA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001928154.7 | XP_001928189.3 | 840 | 83 | ||
| XM_005659540.3 | XP_005659597.1 | 1643 | 83 | |||
| XM_021073474.1 | XP_020929133.1 | 1009 | 83 | |||
| XM_021073481.1 | XP_020929140.1 | 11853 | 83 | |||
| GenBank mRNA | AK347139.1 | - | 3 - 4 | 768 | 83 | |
Set Membership: |
|