Search
Search
| Gene Symbol: MTHFR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003127575.4 | XP_003127623.1 | 1127 | 112 | ||
| XM_021095342.1 | XP_020951001.1 | 1130 | 112 | |||
| GenBank mRNA | AY046558.1 | - | 1 - 2 | 109 | 111 | |
Set Membership: |
|