Search
Search
| Gene Symbol: KPNA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013999108.2 | XP_013854562.2 | 1567 | 113 | ||
| XM_021095806.1 | XP_020951465.1 | 1845 | 113 | |||
| GenBank mRNA | AY010068.2 | - | 3 | 344 | 113 | |
Set Membership: |
|