Search
Search
| Gene Symbol: KIT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044525.1 | NP_001037990.1 | 21 | 2855 | 91 | |
| GenBank mRNA | AB250963.1 | - | 21 | 2855 | 91 | |
| AJ223228.1 | - | 21 | 2894 | 91 | ||
| AJ223229.1 | - | 21 | 2894 | 91 | ||
| AJ223230.1 | - | 21 | 2894 | 91 | ||
| AJ223231.1 | - | 20 | 2771 | 91 | ||
| AK238625.1 | - | 7 | 700 | 91 | ||
| AK347535.1 | - | 21 | 2887 | 91 | ||
| AK350384.1 | - | 10 | 1004 | 91 | ||
| AK398680.1 | - | 11 | 1166 | 91 | ||
| DQ288943.1 | - | 21 | 2916 | 91 | ||
| DQ288944.1 | - | 21 | 2916 | 91 | ||
| FJ938289.1 | - | 21 | 2867 | 91 | ||
| GU356633.1 | - | 20 | 2657 | 91 | ||
| JQ839265.1 | - | 21 | 2826 | 91 | ||
| JQ839266.1 | - | 21 | 2826 | 91 | ||
Set Membership: |
|