Search
Search
| Gene Symbol: AQP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110424.1 | NP_001103894.1 | 1 - 2 | 371 | 105 | |
| GenBank mRNA | AK237974.1 | - | 1 - 2 | 492 | 105 | |
| EU192130.1 | - | 1 - 2 | 371 | 105 | ||
Set Membership: |
|