Search
Search
| Gene Symbol: SIRT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114270.2 | NP_001107742.2 | 2 - 3 | 557 | 88 | |
| GenBank mRNA | AK232110.1 | - | 2 - 3 | 557 | 88 | |
| EU090798.1 | - | 1 - 2 | 323 | 88 | ||
| EU357900.1 | - | 2 - 3 | 515 | 88 | ||
Set Membership: |
|