Search
Search
| Gene Symbol: RPN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213941.1 | NP_999106.1 | 12 - 13 | 1533 | 81 | |
| XM_021077216.1 | XP_020932875.1 | 1525 | 81 | |||
| XM_021077217.1 | XP_020932876.1 | 1525 | 81 | |||
| XM_021077219.1 | XP_020932878.1 | 1824 | 81 | |||
| XM_021077220.1 | XP_020932879.1 | 1805 | 81 | |||
| GenBank mRNA | AJ293583.1 | - | 12 - 13 | 1533 | 81 | |
| AK343289.1 | - | 12 - 13 | 1748 | 81 | ||
| AK391510.1 | - | 6 - 7 | 754 | 81 | ||
| AK392184.1 | - | 12 - 13 | 1515 | 81 | ||
| AK399150.1 | - | 12 - 13 | 1797 | 81 | ||
| AY609499.1 | - | 12 - 13 | 1692 | 81 | ||
Set Membership: |
|