Search
Search
| Gene Symbol: CRNN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159659.1 | NP_001153131.1 | 2 - 3 | 200 | 104 | |
| XM_013997109.2 | XP_013852563.1 | 266 | 104 | |||
| XM_013997110.2 | XP_013852564.1 | 279 | 104 | |||
Set Membership: |
|