Search
Search
| Gene Symbol: ECHS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190175.1 | NP_001177104.1 | 2 - 3 | 373 | 73 | |
| GenBank mRNA | AK232518.1 | - | 2 - 3 | 373 | 73 | |
| AK237861.1 | - | 2 - 3 | 321 | 73 | ||
| AK346255.1 | - | 1 - 2 | 336 | 73 | ||
| AK346919.1 | - | 2 - 3 | 301 | 73 | ||
| AK389542.1 | - | 2 - 3 | 527 | 73 | ||
| AK392778.1 | - | 2 - 3 | 294 | 73 | ||
Set Membership: |
|