Search
Search
| Gene Symbol: ALG10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244234.1 | NP_001231163.1 | 2 - 3 | 515 | 88 | |
| XM_013997746.2 | XP_013853200.1 | 331 | 88 | |||
| GenBank mRNA | AK236309.1 | - | 2 - 3 | 514 | 88 | |
| BW957963.1 | - | 2 - 3 | 490 | 88 | ||
Set Membership: |
|