Search
Search
| Gene Symbol: RB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285967.1 | NP_001272896.1 | 26 - 27 | 2881 | 69 | |
| XM_013992198.2 | XP_013847652.2 | 2853 | 69 | |||
| GenBank mRNA | JX099502.1 | - | 26 - 27 | 2881 | 69 | |
Set Membership: |
|