Search
Search
| Gene Symbol: LAMC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021063797.1 | XP_020919456.1 | 4712 | 92 | ||
| XM_021063798.1 | XP_020919457.1 | 3054 | 92 | |||
| XM_021063799.1 | XP_020919458.1 | 3095 | 92 | |||
| XM_021063800.1 | XP_020919459.1 | 3748 | 92 | |||
| XM_021063801.1 | XP_020919460.1 | 2979 | 92 | |||
| XM_021063802.1 | XP_020919461.1 | 2796 | 92 | |||
| XM_021063803.1 | XP_020919462.1 | 970 | 92 | |||
Set Membership: |
|