Search
Search
| Gene Symbol: AK9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021091041.1 | XP_020946700.1 | 2205 | 189 | ||
| XM_021091043.1 | XP_020946702.1 | 2253 | 189 | |||
| XM_021091050.1 | XP_020946709.1 | 2100 | 189 | |||
| XM_021091055.1 | XP_020946714.1 | 2253 | 189 | |||
| XM_021091063.1 | XP_020946722.1 | 2383 | 189 | |||
Set Membership: |
|