Search
Search
| Gene Symbol: FUBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003127917.5 | XP_003127965.2 | 1818 | 73 | ||
| XM_005665367.3 | XP_005665424.1 | 1818 | 73 | |||
| XM_013999331.2 | XP_013854785.1 | 1818 | 73 | |||
| XM_013999334.2 | XP_013854788.1 | 1818 | 73 | |||
| XM_013999338.2 | XP_013854792.1 | 1752 | 73 | |||
| XM_021096426.1 | XP_020952085.1 | 1815 | 73 | |||
| XM_021096428.1 | XP_020952087.1 | 1818 | 73 | |||
| XM_021096429.1 | XP_020952088.1 | 1755 | 73 | |||
| XM_021096430.1 | XP_020952089.1 | 1815 | 73 | |||
| XM_021096431.1 | XP_020952090.1 | 1752 | 73 | |||
| XM_021096432.1 | XP_020952091.1 | 1755 | 73 | |||
| XM_021096433.1 | XP_020952092.1 | 1815 | 73 | |||
| XM_021096434.1 | XP_020952093.1 | 1752 | 73 | |||
| XM_021096436.1 | XP_020952095.1 | 1755 | 73 | |||
| XM_021096437.1 | XP_020952096.1 | 1755 | 73 | |||
| XM_021096438.1 | XP_020952097.1 | 1752 | 73 | |||
Set Membership: |
|