Search
Search
| Gene Symbol: GASK1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003482392.4 | XP_003482440.1 | 206 | 106 | ||
| XM_005666658.3 | XP_005666715.1 | 208 | 106 | |||
| GenBank mRNA | AK233018.1 | - | 1 - 2 | 173 | 106 | |
| AK396463.1 | - | 1 - 2 | 173 | 106 | ||
Set Membership: |
|