Search
Search

OSBPL1A| Gene Symbol: OSBPL1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003356381.3 | XP_003356429.1 | 681 | 72 | ||
| XM_003356382.4 | XP_003356430.1 | 806 | 72 | |||
| XM_005665323.3 | XP_005665380.3 | 1639 | 72 | |||
| XM_013999263.2 | XP_013854717.2 | 1828 | 72 | |||
| XM_021096188.1 | XP_020951847.1 | 2062 | 72 | |||
| XM_021096189.1 | XP_020951848.1 | 1729 | 72 | |||
| XM_021096190.1 | XP_020951849.1 | 1967 | 72 | |||
| GenBank mRNA | AK233522.1 | - | 4 - 5 | 514 | 72 | |
| AK235755.1 | - | 4 - 5 | 484 | 72 | ||
| AK394099.1 | - | 5 - 6 | 797 | 72 | ||
Set Membership: |
|