Search
Search
| Gene Symbol: HECTD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013978136.2 | XP_013833590.2 | 2036 | 70 | ||
| XM_021099877.1 | XP_020955536.1 | 2036 | 70 | |||
| XM_021099878.1 | XP_020955537.1 | 2036 | 70 | |||
| XM_021099879.1 | XP_020955538.1 | 2036 | 70 | |||
| XM_021099880.1 | XP_020955539.1 | 2036 | 70 | |||
| XM_021099881.1 | XP_020955540.1 | 2036 | 70 | |||
| XM_021099882.1 | XP_020955541.1 | 2036 | 70 | |||
| XM_021099883.1 | XP_020955542.1 | 2036 | 70 | |||
| XM_021099884.1 | XP_020955543.1 | 2036 | 70 | |||
Set Membership: |
|