Search
Search
| Gene Symbol: MYLK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013977395.2 | XP_013832849.2 | 2208 | 79 | ||
| XM_021100109.1 | XP_020955768.1 | 1879 | 79 | |||
| XM_021100110.1 | XP_020955769.1 | 1843 | 79 | |||
| XM_021100111.1 | XP_020955770.1 | 1884 | 79 | |||
| XM_021100112.1 | XP_020955771.1 | 1150 | 79 | |||
| XM_021100113.1 | XP_020955772.1 | 1468 | 79 | |||
Set Membership: |
|