Search
Search

CSNK1G3| Gene Symbol: CSNK1G3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021084688.1 | XP_020940347.1 | 774 | 126 | ||
| XM_021084690.1 | XP_020940349.1 | 834 | 126 | |||
| XM_021084691.1 | XP_020940350.1 | 440 | 126 | |||
| XM_021084692.1 | XP_020940351.1 | 774 | 126 | |||
| XM_021084693.1 | XP_020940352.1 | 774 | 126 | |||
| XM_021084694.1 | XP_020940353.1 | 774 | 126 | |||
| XM_021084695.1 | XP_020940354.1 | 774 | 126 | |||
| XM_021084696.1 | XP_020940355.1 | 873 | 126 | |||
| XM_021084697.1 | XP_020940356.1 | 885 | 126 | |||
| XM_021084698.1 | XP_020940357.1 | 866 | 126 | |||
| XM_021084699.1 | XP_020940358.1 | 885 | 126 | |||
| XM_021084700.1 | XP_020940359.1 | 868 | 126 | |||
Set Membership: |
|