Search
Search
| Gene Symbol: RCBTB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005668401.3 | XP_005668458.1 | 1576 | 81 | ||
| XM_013980497.2 | XP_013835951.1 | 1661 | 81 | |||
| XM_013980504.2 | XP_013835958.1 | 1511 | 81 | |||
| XM_021065307.1 | XP_020920966.1 | 1573 | 81 | |||
| XM_021065308.1 | XP_020920967.1 | 1491 | 81 | |||
| XM_021065309.1 | XP_020920968.1 | 1488 | 81 | |||
| XM_021065310.1 | XP_020920969.1 | 1676 | 81 | |||
| XM_021065311.1 | XP_020920970.1 | 1596 | 81 | |||
Set Membership: |
|