Search
Search
| Gene Symbol: KCNQ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021077758.1 | XP_020933417.1 | 1752 | 71 | ||
| XM_021077760.1 | XP_020933419.1 | 1749 | 71 | |||
| XM_021077761.1 | XP_020933420.1 | 1723 | 71 | |||
| XM_021077762.1 | XP_020933421.1 | 1716 | 71 | |||
| XM_021077763.1 | XP_020933422.1 | 1697 | 71 | |||
| XM_021077764.1 | XP_020933423.1 | 1695 | 71 | |||
| XM_021077765.1 | XP_020933424.1 | 1668 | 71 | |||
| XM_021077766.1 | XP_020933425.1 | 1662 | 71 | |||
| XM_021077767.1 | XP_020933426.1 | 1752 | 71 | |||
| XM_021077768.1 | XP_020933427.1 | 1749 | 71 | |||
| XM_021077769.1 | XP_020933428.1 | 1633 | 71 | |||
| XM_021077770.1 | XP_020933429.1 | 1722 | 71 | |||
| XM_021077771.1 | XP_020933430.1 | 1697 | 71 | |||
| XM_021077772.1 | XP_020933431.1 | 1694 | 71 | |||
| XM_021077773.1 | XP_020933432.1 | 1668 | 71 | |||
| XM_021077774.1 | XP_020933433.1 | 1661 | 71 | |||
| XM_021077775.1 | XP_020933434.1 | 1659 | 71 | |||
Set Membership: |
|